Assay Details
Target Gene Details
Entrez Gene ID: | 60561 |
Gene Name: | RAD50 interactor 1 |
Gene Aliases: |
RINT-1 |
Location: |
Chr.7:105532085-105567688 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RINT1 | NM_021930.4 | NP_068749.3 | ||
| XM_017012496.1 | XP_016867985.1 | |||
| XM_017012497.1 | XP_016867986.1 | |||
| XM_017012498.1 | XP_016867987.1 | |||
| AF317622.1 | AAG42101.1 | |||
| AK055508.1 | ||||
| AK308126.1 | ||||
| BC068483.1 | AAH68483.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1023564 | Chr.7:105378037 - 105542551 on Build GRCh38 | Gain |
|
| dgv6547n100 | Chr.7:105166760 - 105542551 on Build GRCh38 | Gain |
|
| nsv464668 | Chr.7:105448246 - 105568045 on Build GRCh38 | Gain |
|
| esv3614417 | Chr.7:105370987 - 105549944 on Build GRCh38 | Gain |
|
| esv2759549 | Chr.7:105117803 - 105568045 on Build GRCh38 | Gain |
|
| esv3614427 | Chr.7:105529416 - 105544030 on Build GRCh38 | Gain |
|
| esv3891174 | Chr.7:105387199 - 105542548 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map