Assay Details
Target Gene Details
Entrez Gene ID: | 57414 |
Gene Name: | rhomboid domain containing 2 |
Gene Aliases: |
NPD007, RHBDL7 |
Location: |
Chr.7:75878999-75888926 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RHBDD2 | NM_001040456.1 | NP_001035546.1 | ||
| NM_001040457.1 | NP_001035547.1 | |||
| XM_005250511.4 | XP_005250568.1 | |||
| AF226732.1 | AAG09733.1 | |||
| AK026590.1 | BAB15503.1 | |||
| BC006234.2 | AAH06234.2 | |||
| BC069017.1 | AAH69017.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528969 | Chr.7:75074053 - 76920728 on Build GRCh38 | Gain |
|
| dgv120n16 | Chr.7:75440769 - 76641919 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map