Assay Details
Target Gene Details
Entrez Gene ID: | 222901 |
Gene Name: | ribosomal protein L23 pseudogene 8 |
Gene Aliases: |
RPL23_4_790 |
Location: |
Chr.7:20827298-20827820 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPL23P8 | NR_026673.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1030726 | Chr.7:20466472 - 21047100 on Build GRCh38 | Gain |
|

Genomic Map