Assay Details
Target Gene Details
Entrez Gene ID: | 27342 |
Gene Name: | RAB guanine nucleotide exchange factor 1 |
Gene Aliases: |
RABEX5, RAP1, rabex-5 |
Location: |
Chr.7:66682091-66811464 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RABGEF1 | NM_001287060.1 | NP_001273989.1 | ||
| NM_001287061.1 | NP_001273990.1 | |||
| NM_001287062.1 | NP_001273991.1 | |||
| NM_014504.2 | NP_055319.1 | |||
| AJ250042.1 | CAB57359.1 | |||
| AK001702.1 | ||||
| AK056864.1 | ||||
| AK223329.1 | BAD97049.1 | |||
| AK290678.1 | ||||
| AK303006.1 | ||||
| BC015330.1 | AAH15330.1 | |||
| BT007107.1 | AAP35771.1 | |||
| DQ230533.1 | ABA64473.1 | |||
| DQ230534.1 | ABA64474.1 | |||
| DQ230535.1 | ABA64475.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3613520 | Chr.7:66748771 - 66896312 on Build GRCh38 | Gain |
|
| esv3613521 | Chr.7:66755218 - 66838078 on Build GRCh38 | Gain |
|
| nsv5773 | Chr.7:66770168 - 66801796 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map