Assay Details
Target Gene Details
Entrez Gene ID: | 346689 |
Gene Name: | killer cell lectin like receptor G2 |
Gene Aliases: |
CLEC15B |
Location: |
Chr.7:139426931-139483716 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KLRG2 | XM_011516141.2 | 4 | 1474 | XP_011514443.1 |
Target Gene Details
Entrez Gene ID: | 100129148 |
Gene Name: | uncharacterized LOC100129148 |
Gene Aliases: |
- |
Location: |
Chr.7:139417462-139427526 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100129148 | NR_033999.1 | |||
| AI811258.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2735212 | Chr.7:138060002 - 140415947 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map