Assay Details
Target Gene Details
Entrez Gene ID: | 3574 |
Gene Name: | interleukin 7 |
Gene Aliases: |
IL-7 |
Location: |
Chr.8:78675870-78805523 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL7 | XM_011517522.2 | XP_011515824.1 | ||
| XM_011517523.2 | XP_011515825.1 |
Target Gene Details
Entrez Gene ID: | 101241902 |
Gene Name: | chromosome 4 open reading frame 46 pseudogene |
Gene Aliases: |
- |
Location: |
Chr.8:78722847-78724374 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101241902 | NR_077236.1 | 1 | 915 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3891420 | Chr.8:78710128 - 78731403 on Build GRCh38 | Loss |
|
| esv2759622 | Chr.8:78709999 - 78876112 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs118001357] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map