Assay Details
Target Gene Details
Entrez Gene ID: | 101927543 |
Gene Name: | uncharacterized LOC101927543 |
Gene Aliases: |
- |
Location: |
Chr.8:120761253-120776832 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101927543 | NR_125419.1 | |||
| AK057448.1 |
Target Gene Details
Entrez Gene ID: | 6641 |
Gene Name: | syntrophin beta 1 |
Gene Aliases: |
59-DAP, A1B, BSYN2, DAPA1B, SNT2, SNT2B1, TIP-43 |
Location: |
Chr.8:120535745-120812069 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SNTB1 | NM_021021.3 | NP_066301.1 | ||
| XM_011517239.1 | XP_011515541.1 | |||
| AF028828.1 | AAB84253.1 | |||
| AK026095.1 | ||||
| AK292655.1 | ||||
| BC098573.1 | AAH98573.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1016172 | Chr.8:120667350 - 120917676 on Build GRCh38 | Loss |
|
| esv2759640 | Chr.8:120753461 - 120816679 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map