Assay Details
Target Gene Details
Entrez Gene ID: | 102723729 |
Gene Name: | uncharacterized LOC102723729 |
Gene Aliases: |
- |
Location: |
Chr.8:41534305-41578368 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC102723729 | NR_125824.1 | |||
| AI702202.1 | ||||
| BE326543.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831299 | Chr.8:41448617 - 41692486 on Build GRCh38 | Loss |
|
| esv3576565 | Chr.8:41064045 - 41790840 on Build GRCh38 | Gain |
|
| dgv2070e212 | Chr.8:41543262 - 41552999 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74909169] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map