Assay Details
Target Gene Details
Entrez Gene ID: | 401466 |
Gene Name: | chromosome 8 open reading frame 59 |
Gene Aliases: |
- |
Location: |
Chr.8:85214053-85220422 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C8orf59 | NM_001099670.2 | NP_001093140.1 | ||
| NM_001099671.2 | NP_001093141.1 | |||
| NM_001099672.2 | NP_001093142.1 | |||
| NM_001099673.2 | NP_001093143.1 | |||
| NM_001293320.1 | NP_001280249.1 | |||
| XM_011517528.2 | XP_011515830.1 | |||
| BC146983.1 | ||||
| BC146988.1 | ||||
| BC157825.1 | ||||
| BM785759.1 | ||||
| CB119146.1 | ||||
| CN280666.1 | ||||
| CN481353.1 | ||||
| DB094478.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv483126 | Chr.8:83687766 - 85887771 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map