Assay Details
Target Gene Details
Entrez Gene ID: | 27067 |
Gene Name: | staufen double-stranded RNA binding protein 2 |
Gene Aliases: |
39K2, 39K3 |
Location: |
Chr.8:73420369-73747708 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STAU2 | NM_001164380.1 | NP_001157852.1 | ||
| NM_001164381.1 | NP_001157853.1 | |||
| NM_001164382.1 | NP_001157854.1 | |||
| NM_001164383.1 | NP_001157855.1 | |||
| AF459097.1 | AAN37926.1 | |||
| AF459098.1 | AAN37927.1 | |||
| AK002152.1 | BAA92111.1 | |||
| AK293496.1 | ||||
| AK303104.1 | ||||
| BC008369.1 | AAH08369.1 |
Target Gene Details
Entrez Gene ID: | 100128126 |
Gene Name: | STAU2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.8:73420074-73441518 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STAU2-AS1 | NR_038406.1 | |||
| AK056720.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1020902 | Chr.8:73413975 - 73464215 on Build GRCh38 | Loss |
|
| esv2737154 | Chr.8:71825978 - 73469084 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map