Assay Details
Target Gene Details
Entrez Gene ID: | 59339 |
Gene Name: | pleckstrin homology domain containing A2 |
Gene Aliases: |
TAPP2 |
Location: |
Chr.8:38901235-38973912 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLEKHA2 | NM_021623.1 | 1 | 123 | NP_067636.1 |
| XM_017013718.1 | XP_016869207.1 | |||
| AF286164.1 | 1 | 47 | AAG15201.1 | |
| AK098787.1 | 1 | 123 | ||
| AK291842.1 | 1 | 67 | ||
| AK296068.1 | 1 | 79 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv610950 | Chr.8:38897012 - 38940631 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map