Assay Details
Target Gene Details
Entrez Gene ID: | 157285 |
Gene Name: | homolog of rat pragma of Rnd2 |
Gene Aliases: |
PRAGMIN |
Location: |
Chr.8:8317731-8386444 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SGK223 | NM_001080826.2 | NP_001074295.2 | ||
| XM_005272369.4 | XP_005272426.2 | |||
| XM_005272370.4 | XP_005272427.2 | |||
| AK122582.1 | BAC56923.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv610154 | Chr.8:8244119 - 8470591 on Build GRCh38 | Loss |
|
| esv2764098 | Chr.8:7102311 - 8743201 on Build GRCh38 | Gain+Loss |
|
| nsv610153 | Chr.8:8240557 - 8597021 on Build GRCh38 | Loss |
|
| esv2759589 | Chr.8:6874198 - 8449233 on Build GRCh38 | Gain+Loss |
|
| esv3616132 | Chr.8:8331077 - 8335018 on Build GRCh38 | Loss |
|
| nsv610156 | Chr.8:8305721 - 8349823 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74609549] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map