Assay Details
Target Gene Details
Entrez Gene ID: | 137886 |
Gene Name: | UBX domain protein 2B |
Gene Aliases: |
p37 |
Location: |
Chr.8:58411264-58451508 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBXN2B | NM_001077619.1 | NP_001071087.1 | ||
| XM_005251165.2 | XP_005251222.1 | |||
| AK126300.1 | ||||
| AL831990.2 | ||||
| AL833075.2 | ||||
| BC113645.1 | ||||
| BX647876.2 |
More Information
Set Membership: |
|

Genomic Map