Assay Details
Target Gene Details
Entrez Gene ID: | 100132891 |
Gene Name: | MSC antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.8:71843123-72056312 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MSC-AS1 | NR_033651.1 | |||
| NR_033652.1 | ||||
| AK123315.1 | ||||
| AK304427.1 | ||||
| BX647470.1 | ||||
| DB161440.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831353 | Chr.8:71836557 - 72032198 on Build GRCh38 | Loss |
|
| esv2737154 | Chr.8:71825978 - 73469084 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs77678832] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map