Assay Details
Target Gene Details
Entrez Gene ID: | 4023 |
Gene Name: | lipoprotein lipase |
Gene Aliases: |
HDLCQ11, LIPD |
Location: |
Chr.8:19939071-19967259 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LPL | NM_000237.2 | NP_000228.1 | ||
| AK092286.1 | ||||
| AK222464.1 | BAD96184.1 | |||
| AK290290.1 | ||||
| AK297164.1 | ||||
| AK312311.1 | ||||
| BC011353.1 | AAH11353.1 | |||
| BT006726.1 | AAP35372.1 | |||
| CR457054.1 | CAG33335.1 | |||
| M15856.1 | AAB59536.1 | |||
| X14390.1 | CAA32564.1 | |||
| X54516.1 | CAA38372.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3616553 | Chr.8:19911549 - 20084901 on Build GRCh38 | Gain |
|
| esv32767 | Chr.8:19882678 - 20015571 on Build GRCh38 | Loss |
|
| nsv610759 | Chr.8:19704486 - 20211331 on Build GRCh38 | Gain |
|
| nsv522185 | Chr.8:19097292 - 20497802 on Build GRCh38 | Gain |
|
| esv3891372 | Chr.8:19744844 - 20118728 on Build GRCh38 | Gain |
|
| nsv1033511 | Chr.8:19893788 - 20767580 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map