Assay Details
Target Gene Details
Entrez Gene ID: | 59339 |
Gene Name: | pleckstrin homology domain containing A2 |
Gene Aliases: |
TAPP2 |
Location: |
Chr.8:38901235-38973912 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLEKHA2 | NM_021623.1 | NP_067636.1 | ||
| XM_011544605.2 | XP_011542907.1 | |||
| XM_011544607.2 | XP_011542909.1 | |||
| XM_017013718.1 | XP_016869207.1 | |||
| XM_017013719.1 | XP_016869208.1 | |||
| XM_017013720.1 | XP_016869209.1 | |||
| AF286164.1 | AAG15201.1 | |||
| AK098787.1 | ||||
| AK291842.1 | ||||
| AK296068.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv610950 | Chr.8:38897012 - 38940631 on Build GRCh38 | Loss |
|
| esv3616935 | Chr.8:38903921 - 38908617 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map