Assay Details
Target Gene Details
Entrez Gene ID: | 6101 |
Gene Name: | retinitis pigmentosa 1 (autosomal dominant) |
Gene Aliases: |
DCDC4A, ORP1 |
Location: |
Chr.8:54509448-54630834 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RP1 | XM_017013721.1 | XP_016869210.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509267 | Chr.8:54511442 - 54559433 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map