Assay Details
Target Gene Details
Entrez Gene ID: | 83656 |
Gene Name: | FAM167A antisense RNA 1 |
Gene Aliases: |
C8orf12 |
Location: |
Chr.8:11368402-11438657 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM167A-AS1 | NR_026814.1 | |||
| AJ301563.1 | CAC82739.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34097 | Chr.8:11319863 - 11632385 on Build GRCh38 | Loss |
|
| nsv831233 | Chr.8:11189345 - 11376875 on Build GRCh38 | Gain |
|
| nsv1033312 | Chr.8:11269291 - 11394661 on Build GRCh38 | Loss |
|

Genomic Map