Assay Details
Target Gene Details
Entrez Gene ID: | 137814 |
Gene Name: | NK2 homeobox 6 |
Gene Aliases: |
CSX2, CTHM, NKX2F, NKX4-2 |
Location: |
Chr.8:23702451-23706598 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NKX2-6 | NM_001136271.2 | NP_001129743.2 | ||
| CN272646.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831269 | Chr.8:23661667 - 23849831 on Build GRCh38 | Gain |
|
| nsv1028341 | Chr.8:23550380 - 24287905 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map