Assay Details
Target Gene Details
Entrez Gene ID: | 4067 |
Gene Name: | LYN proto-oncogene, Src family tyrosine kinase |
Gene Aliases: |
JTK8, p53Lyn, p56Lyn |
Location: |
Chr.8:55877305-56012447 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LYN | NM_001111097.2 | NP_001104567.1 | ||
| NM_002350.3 | NP_002341.1 | |||
| XM_011517529.2 | XP_011515831.1 | |||
| XM_017013415.1 | XP_016868904.1 | |||
| AK290494.1 | ||||
| BC059394.1 | AAH59394.1 | |||
| BC068551.1 | AAH68551.1 | |||
| BC126456.1 | ||||
| BC126458.1 | ||||
| CB990993.1 | ||||
| M16038.1 | AAA59540.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3617285 | Chr.8:55864913 - 55911860 on Build GRCh38 | Gain |
|
| dgv889n27 | Chr.8:55799154 - 55963660 on Build GRCh38 | Gain |
|
| esv3617283 | Chr.8:55804919 - 55968939 on Build GRCh38 | Gain |
|
| dgv459n21 | Chr.8:55676869 - 56030580 on Build GRCh38 | Gain |
|
| esv3891398 | Chr.8:55878090 - 56029138 on Build GRCh38 | Gain |
|
| esv2752257 | Chr.8:55867087 - 55969887 on Build GRCh38 | Gain |
|
| nsv527341 | Chr.8:55679732 - 55895699 on Build GRCh38 | Gain |
|
| dgv7228n100 | Chr.8:55806899 - 55964201 on Build GRCh38 | Gain |
|
| dgv12173n54 | Chr.8:55794435 - 55963660 on Build GRCh38 | Gain |
|
| nsv611393 | Chr.8:55763923 - 55993509 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs144157801] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map