Assay Details
Target Gene Details
Entrez Gene ID: | 10584 |
Gene Name: | collectin subfamily member 10 |
Gene Aliases: |
CL-34, CLL1 |
Location: |
Chr.8:118952167-119108455 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COLEC10 | NM_001324095.1 | NP_001311024.1 | ||
| XM_005250756.3 | XP_005250813.1 |
Target Gene Details
Entrez Gene ID: | 101927513 |
Gene Name: | uncharacterized LOC101927513 |
Gene Aliases: |
- |
Location: |
Chr.8:119062942-119068782 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101927513 | NR_134297.1 | 2 | 1232 | |
| DB519943.1 | 2 | 284 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759639 | Chr.8:118931070 - 119263510 on Build GRCh38 | Gain |
|
| esv3891471 | Chr.8:119015632 - 119068453 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map