Assay Details
Target Gene Details
Entrez Gene ID: | 8822 |
Gene Name: | fibroblast growth factor 17 |
Gene Aliases: |
FGF-13, FGF-17, HH20 |
Location: |
Chr.8:22039708-22048809 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FGF17 | XM_011544683.1 | 1 | 1348 | XP_011542985.1 |
| XM_011544684.2 | 2 | 648 | XP_011542986.1 | |
| XM_011544685.1 | 1 | 1348 | XP_011542987.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528590 | Chr.8:22034428 - 22168131 on Build GRCh38 | Loss |
|
| nsv610764 | Chr.8:22031342 - 22094715 on Build GRCh38 | Loss |
|
| nsv951112 | Chr.8:22023690 - 22135287 on Build GRCh38 | Deletion |
|
| nsv831263 | Chr.8:22031088 - 22184413 on Build GRCh38 | Loss |
|
| esv2759604 | Chr.8:20675533 - 23254096 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map