Assay Details
Target Gene Details
Entrez Gene ID: | 6867 |
Gene Name: | transforming acidic coiled-coil containing protein 1 |
Gene Aliases: |
Ga55 |
Location: |
Chr.8:38728186-38853028 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TACC1 | NM_001146216.2 | NP_001139688.1 | ||
| XM_005273625.4 | XP_005273682.1 | |||
| XM_005273629.1 | XP_005273686.1 | |||
| XM_005273630.1 | XP_005273687.1 | |||
| XM_011544632.2 | XP_011542934.1 | |||
| XM_017013767.1 | XP_016869256.1 | |||
| XM_017013769.1 | XP_016869258.1 | |||
| XM_017013771.1 | XP_016869260.1 | |||
| XM_017013773.1 | XP_016869262.1 | |||
| XM_017013775.1 | XP_016869264.1 | |||
| XM_017013777.1 | XP_016869266.1 | |||
| AK124297.1 | ||||
| AK295071.1 | ||||
| AK303596.1 | ||||
| AK304507.1 | ||||
| AK304774.1 | ||||
| AY072873.2 | AAL62460.2 | |||
| AY072876.2 | AAL62463.2 | |||
| BC041391.1 | AAH41391.1 | |||
| BX647696.1 | CAI46025.1 | |||
| DA553264.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831296 | Chr.8:38663712 - 38800294 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map