Assay Details
Target Gene Details
Entrez Gene ID: | 445577 |
Gene Name: | chromosome 9 open reading frame 129 |
Gene Aliases: |
bA165J3.3 |
Location: |
Chr.9:93318199-93346414 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C9orf129 | NM_001098808.1 | NP_001092278.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv972783 | Chr.9:93342714 - 93346481 on Build GRCh38 | Duplication |
|

Genomic Map