Assay Details
Target Gene Details
Entrez Gene ID: | 5524 |
Gene Name: | protein phosphatase 2 phosphatase activator |
Gene Aliases: |
PP2A, PPP2R4, PR53 |
Location: |
Chr.9:129110949-129148946 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv818725 | Chr.9:129026890 - 129176634 on Build GRCh38 | Gain |
|
| nsv831729 | Chr.9:128973042 - 129158330 on Build GRCh38 | Loss |
|
| nsv509323 | Chr.9:128991062 - 129129399 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map