Assay Details
Target Gene Details
Entrez Gene ID: | 100286938 |
Gene Name: | VPS13A antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.9:77176756-77177917 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VPS13A-AS1 | NR_026668.2 | |||
| AA626167.1 | ||||
| DB509755.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv7682n100 | Chr.9:77154478 - 77180761 on Build GRCh38 | Gain |
|

Genomic Map