Assay Details
Target Gene Details
Entrez Gene ID: | 6456 |
Gene Name: | SH3 domain containing GRB2 like 2, endophilin A1 |
Gene Aliases: |
CNSA2, EEN-B1, SH3D2A, SH3P4 |
Location: |
Chr.9:17578954-17797129 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SH3GL2 | NM_003026.3 | NP_003017.1 | ||
| AB206840.1 | BAE44459.1 | |||
| AF036268.1 | AAC04764.1 | |||
| AK057744.1 | ||||
| AK293546.1 | ||||
| AK293592.1 | ||||
| AK312400.1 | ||||
| BC032825.2 | AAH32825.1 | |||
| BT019682.1 | AAV38488.1 | |||
| CR542086.1 | CAG46883.1 | |||
| CR542102.1 | CAG46899.1 | |||
| DC309737.1 | ||||
| X99657.1 | CAA67971.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv7466n100 | Chr.9:17577143 - 17636676 on Build GRCh38 | Gain |
|
| esv3573159 | Chr.9:17506784 - 17662496 on Build GRCh38 | Loss |
|
| esv2422265 | Chr.9:17396713 - 17598189 on Build GRCh38 | Duplication |
|
| nsv466269 | Chr.9:17165700 - 17710409 on Build GRCh38 | Gain |
|
| nsv613693 | Chr.9:17147193 - 17710409 on Build GRCh38 | Gain |
|
| nsv517380 | Chr.9:17564269 - 17633234 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map