Assay Details
Target Gene Details
Entrez Gene ID: | 730112 |
Gene Name: | family with sequence similarity 166 member B |
Gene Aliases: |
- |
Location: |
Chr.9:35561830-35563899 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM166B | NM_001099951.3 | NP_001093421.1 | ||
| NM_001164310.2 | NP_001157782.1 | |||
| NM_001287238.1 | NP_001274167.1 | |||
| NM_001287239.1 | NP_001274168.1 | |||
| XM_011518028.2 | 2 | 194 | XP_011516330.1 | |
| BC129999.1 | ||||
| BC146928.1 | ||||
| BC146933.1 | ||||
| BC171840.1 | ||||
| BC171851.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3620379 | Chr.9:35504377 - 35568617 on Build GRCh38 | Gain |
|
| nsv527028 | Chr.9:35557209 - 35574612 on Build GRCh38 | Loss |
|
| nsv951701 | Chr.9:35555404 - 35563903 on Build GRCh38 | Deletion |
|

Genomic Map