Assay Details
Target Gene Details
Entrez Gene ID: | 375757 |
Gene Name: | SWI5 homologous recombination repair protein |
Gene Aliases: |
C9orf119, SAE3 |
Location: |
Chr.9:128275379-128288989 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SWI5 | NM_001040011.1 | 5 | 751 | NP_001035100.1 |
| NM_001318089.1 | 5 | 631 | NP_001305018.1 | |
| NM_001318092.1 | 5 | 492 | NP_001305021.1 | |
| BC029911.1 | 5 | 675 | AAH29911.1 | |
| BG541668.1 | 5 | 534 | ||
| CF129530.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831728 | Chr.9:128237994 - 128416514 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map