Assay Details
Target Gene Details
Entrez Gene ID: | 9853 |
Gene Name: | RUN and SH3 domain containing 2 |
Gene Aliases: |
Iporin |
Location: |
Chr.9:35487999-35561898 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RUSC2 | NM_014806.3 | NP_055621.1 | ||
| NR_052015.2 | ||||
| XM_006716896.2 | XP_006716959.1 | |||
| AB002373.2 | ||||
| BC150262.1 | ||||
| DB458531.2 | ||||
| DB460837.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv968610 | Chr.9:35505261 - 35509524 on Build GRCh38 | Duplication |
|
| nsv1030303 | Chr.9:35084357 - 35506646 on Build GRCh38 | Gain |
|
| esv3620379 | Chr.9:35504377 - 35568617 on Build GRCh38 | Gain |
|
| nsv831545 | Chr.9:35327931 - 35507322 on Build GRCh38 | Loss |
|
| esv3620378 | Chr.9:35499806 - 35547677 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map