Assay Details
Target Gene Details
Entrez Gene ID: | 9536 |
Gene Name: | prostaglandin E synthase |
Gene Aliases: |
MGST-IV, MGST1-L1, MGST1L1, MPGES, PGES, PIG12, PP102, PP1294, TP53I12, mPGES-1 |
Location: |
Chr.9:129738336-129777579 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTGES | XM_011519210.2 | XP_011517512.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv23022 | Chr.9:129764226 - 129767505 on Build GRCh38 | Loss |
|
| nsv1046477 | Chr.9:129608081 - 129997996 on Build GRCh38 | Gain |
|
| esv3621839 | Chr.9:129701704 - 129885823 on Build GRCh38 | Loss |
|
| nsv436625 | Chr.9:129762543 - 129768628 on Build GRCh38 | Deletion |
|
| esv3621844 | Chr.9:129764130 - 129768243 on Build GRCh38 | Loss |
|
| nsv6728 | Chr.9:129752333 - 129784235 on Build GRCh38 | Deletion |
|
| dgv1380e199 | Chr.9:129763416 - 129768463 on Build GRCh38 | Deletion |
|
| nsv515126 | Chr.9:129764204 - 129765660 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs112145353] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map