Assay Details
Target Gene Details
Entrez Gene ID: | 84895 |
Gene Name: | mitoguardin 2 |
Gene Aliases: |
C9orf54, FAM73B |
Location: |
Chr.9:129036605-129072082 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIGA2 | NM_032809.2 | NP_116198.2 | ||
| XM_005252282.1 | XP_005252339.1 | |||
| XM_011519117.1 | XP_011517419.1 | |||
| XM_011519118.1 | XP_011517420.1 | |||
| AK074127.1 | BAB84953.1 | |||
| AK075421.1 | BAC11611.1 | |||
| AK301241.1 | ||||
| AK303036.1 | ||||
| BC009114.1 | AAH09114.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv818725 | Chr.9:129026890 - 129176634 on Build GRCh38 | Gain |
|
| nsv831729 | Chr.9:128973042 - 129158330 on Build GRCh38 | Loss |
|
| nsv509323 | Chr.9:128991062 - 129129399 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map