Assay Details
Target Gene Details
Entrez Gene ID: | 23392 |
Gene Name: | KIAA0368 |
Gene Aliases: |
ECM29 |
Location: |
Chr.9:111360693-111484745 on Build GRCh38 |
Assay Gene Location: | Within Exon 53 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KIAA0368 | NM_001080398.1 | 51 | 6695 | NP_001073867.1 |
| XM_005251850.3 | 49 | 6198 | XP_005251907.1 | |
| AB002366.1 | 39 | 4969 | BAA20823.1 | |
| AF052139.1 | 1 | 514 | ||
| AK124590.1 | 2 | 3476 |
More Information
Set Membership: |
|

Genomic Map