Assay Details
Target Gene Details
Entrez Gene ID: | 9853 |
Gene Name: | RUN and SH3 domain containing 2 |
Gene Aliases: |
Iporin |
Location: |
Chr.9:35487999-35561898 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RUSC2 | XM_006716896.2 | 1 | 1701 | XP_006716959.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1030303 | Chr.9:35084357 - 35506646 on Build GRCh38 | Gain |
|
| nsv831545 | Chr.9:35327931 - 35507322 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map