Assay Details
Target Gene Details
Entrez Gene ID: | 445577 |
Gene Name: | chromosome 9 open reading frame 129 |
Gene Aliases: |
bA165J3.3 |
Location: |
Chr.9:93318199-93346414 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C9orf129 | NM_001098808.1 | NP_001092278.1 | ||
| XM_006717117.3 | XP_006717180.1 |

Genomic Map