Assay Details
Target Gene Details
Entrez Gene ID: | 11258 |
Gene Name: | dynactin subunit 3 |
Gene Aliases: |
DCTN-22, DCTN22 |
Location: |
Chr.9:34613545-34620523 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DCTN3 | NM_001281425.1 | NP_001268354.1 | ||
| NM_001281426.1 | NP_001268355.1 | |||
| NM_001281427.1 | NP_001268356.1 | |||
| NM_007234.4 | NP_009165.1 | |||
| NM_024348.3 | NP_077324.1 | |||
| AF082513.1 | AAC61280.1 | |||
| AK091661.1 | 3 | 1109 | BAC03716.1 | |
| AK314730.1 | ||||
| AW953496.1 | ||||
| BC000319.2 | AAH00319.1 | |||
| BC003004.1 | AAH03004.1 | |||
| BC107697.1 | AAI07698.1 | |||
| BG715167.1 | ||||
| BI596981.1 | ||||
| BI837796.1 | ||||
| CN257020.1 | ||||
| CR541889.1 | CAG46687.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3620335 | Chr.9:33871811 - 34738198 on Build GRCh38 | Gain |
|
| nsv1161857 | Chr.9:34537279 - 34669174 on Build GRCh38 | Deletion |
|
| dgv12659n54 | Chr.9:34552439 - 34663141 on Build GRCh38 | Loss |
|
| nsv818699 | Chr.9:34535211 - 34685943 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map