Assay Details
Target Gene Details
Entrez Gene ID: | 138715 |
Gene Name: | AT-rich interaction domain 3C |
Gene Aliases: |
- |
Location: |
Chr.9:34621360-34633112 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARID3C | XM_017014283.1 | XP_016869772.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3620335 | Chr.9:33871811 - 34738198 on Build GRCh38 | Gain |
|
| nsv1161857 | Chr.9:34537279 - 34669174 on Build GRCh38 | Deletion |
|
| dgv12659n54 | Chr.9:34552439 - 34663141 on Build GRCh38 | Loss |
|
| nsv818699 | Chr.9:34535211 - 34685943 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map