Assay Details
Target Gene Details
Entrez Gene ID: | 375743 |
Gene Name: | protein prenyltransferase alpha subunit repeat containing 1 |
Gene Aliases: |
- |
Location: |
Chr.9:69709522-69760050 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTAR1 | NM_001099666.1 | 8 | 3363 | NP_001093136.1 |
| XM_005251976.4 | 7 | 3534 | XP_005252033.1 | |
| XM_005251977.4 | 8 | 3424 | XP_005252034.1 | |
| XM_011518639.1 | 7 | 3408 | XP_011516941.1 | |
| XM_011518640.1 | 8 | 3584 | XP_011516942.1 | |
| AK127511.1 | 2 | 2698 | ||
| AL832683.1 | 5 | 3188 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3620566 | Chr.9:69712247 - 69717695 on Build GRCh38 | Gain |
|
| nsv471309 | Chr.9:69658982 - 69795407 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map