Assay Details
Target Gene Details
Entrez Gene ID: | 445577 |
Gene Name: | chromosome 9 open reading frame 129 |
Gene Aliases: |
bA165J3.3 |
Location: |
Chr.9:93318199-93346414 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C9orf129 | NM_001098808.1 | NP_001092278.1 | ||
| XM_006717117.3 | XP_006717180.1 |
Target Gene Details
Entrez Gene ID: | 65268 |
Gene Name: | WNK lysine deficient protein kinase 2 |
Gene Aliases: |
NY-CO-43, P/OKcl.13, PRKWNK2, SDCCAG43 |
Location: |
Chr.9:93184156-93327581 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WNK2 |
More Information
Additional Information:
For this assay, SNP(s) [rs114548844] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map