Assay Details
Target Gene Details
Entrez Gene ID: | 387715 |
Gene Name: | age-related maculopathy susceptibility 2 |
Gene Aliases: |
ARMD8 |
Location: |
Chr.10:122454663-122457352 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARMS2 | NM_001099667.1 | 2 | 707 | NP_001093137.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3546969 | Chr.10:122457284 - 122457746 on Build GRCh38 | Deletion |
|
| esv28769 | Chr.10:122457239 - 122457839 on Build GRCh38 | Loss |
|
| nsv7580 | Chr.10:122446276 - 122490980 on Build GRCh38 | Deletion |
|
| nsv825594 | Chr.10:122434264 - 122505709 on Build GRCh38 | Gain |
|
| dgv1409n54 | Chr.10:122457266 - 122457796 on Build GRCh38 | Loss |
|
| esv9290 | Chr.10:122457140 - 122457933 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map