Assay Details
Target Gene Details
Entrez Gene ID: | 390010 |
Gene Name: | NK1 homeobox 2 |
Gene Aliases: |
C10orf121, NKX-1.1, bB238F13.2 |
Location: |
Chr.10:124447429-124449981 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NKX1-2 | NM_001146340.1 | NP_001139812.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832012 | Chr.10:124300733 - 124461668 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map