Assay Details
Target Gene Details
Entrez Gene ID: | 387715 |
Gene Name: | age-related maculopathy susceptibility 2 |
Gene Aliases: |
ARMD8 |
Location: |
Chr.10:122454663-122457352 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARMS2 | NM_001099667.1 | NP_001093137.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv7580 | Chr.10:122446276 - 122490980 on Build GRCh38 | Deletion |
|
| nsv825594 | Chr.10:122434264 - 122505709 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map