Assay Details
Target Gene Details
Entrez Gene ID: | 92565 |
Gene Name: | fibronectin type III and ankyrin repeat domains 1 |
Gene Aliases: |
HSD13 |
Location: |
Chr.10:125896539-126009599 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Exon 5 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2761638 | Chr.10:125892202 - 126013242 on Build GRCh38 | Loss |
|
| esv2759788 | Chr.10:125765331 - 126098133 on Build GRCh38 | Gain |
|

Genomic Map