Assay Details
Target Gene Details
Entrez Gene ID: | 439949 |
Gene Name: | PRKCQ antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.10:6580425-6585361 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRKCQ-AS1 | NR_036502.1 | 3 | 844 | |
| NR_036503.1 | 2 | 780 | ||
| AY007155.1 | 2 | 666 | ||
| DB554966.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1047309 | Chr.10:6453621 - 6672098 on Build GRCh38 | Gain |
|
| nsv5698 | Chr.10:6564860 - 6590481 on Build GRCh38 | Insertion |
|
| nsv549912 | Chr.10:6525013 - 6587263 on Build GRCh38 | Gain |
|

Genomic Map