Assay Details
Target Gene Details
Entrez Gene ID: | 387712 |
Gene Name: | enolase family member 4 |
Gene Aliases: |
C10orf134 |
Location: |
Chr.10:116848230-116911803 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ENO4 | NM_001242699.1 | NP_001229628.1 | ||
| XM_005269815.3 | XP_005269872.1 | |||
| XM_006717835.3 | XP_006717898.1 | |||
| XM_006717836.3 | XP_006717899.1 | |||
| XM_011539797.2 | XP_011538099.1 | |||
| XM_011539799.2 | XP_011538101.1 | |||
| XM_017016241.1 | XP_016871730.1 | |||
| BX647301.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs200198481] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map