Assay Details
Target Gene Details
Entrez Gene ID: | 54906 |
Gene Name: | family with sequence similarity 208 member B |
Gene Aliases: |
C10orf18, bA318E3.2 |
Location: |
Chr.10:5684759-5763740 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM208B | NM_001321783.1 | NP_001308712.1 | ||
| NM_001321784.1 | NP_001308713.1 | |||
| NM_001321785.1 | NP_001308714.1 | |||
| NM_017782.4 | NP_060252.4 | |||
| XM_005252474.3 | 1 | 4570 | XP_005252531.1 | |
| XM_005252475.3 | XP_005252532.1 | |||
| XM_005252476.4 | XP_005252533.1 | |||
| XM_005252480.4 | XP_005252537.1 | |||
| XM_005252481.4 | XP_005252538.1 | |||
| XM_011519515.2 | 1 | 4570 | XP_011517817.1 | |
| XM_011519516.2 | 1 | 4570 | XP_011517818.1 | |
| XM_011519517.2 | XP_011517819.1 | |||
| XM_011519518.2 | XP_011517820.1 | |||
| XM_011519519.2 | 1 | 4570 | XP_011517821.1 | |
| XM_011519520.2 | 1 | 4570 | XP_011517822.1 | |
| XM_011519521.2 | XP_011517823.1 | |||
| XM_011519522.2 | XP_011517824.1 | |||
| XM_017016361.1 | 1 | 4570 | XP_016871850.1 | |
| XM_017016362.1 | XP_016871851.1 | |||
| XM_017016363.1 | XP_016871852.1 | |||
| XM_017016364.1 | XP_016871853.1 | |||
| XM_017016365.1 | XP_016871854.1 | |||
| AB621821.1 | ||||
| AL049233.1 | 1 | 2675 | ||
| CR936780.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522415 | Chr.10:5679690 - 5726530 on Build GRCh38 | Gain |
|
| nsv1050663 | Chr.10:5696016 - 5743955 on Build GRCh38 | Loss |
|
| dgv1e49 | Chr.10:5684858 - 6183224 on Build GRCh38 | Loss |
|
| nsv528504 | Chr.10:5697091 - 5712648 on Build GRCh38 | Gain |
|
| esv2759729 | Chr.10:5515061 - 5762817 on Build GRCh38 | Gain |
|

Genomic Map