Assay Details
Target Gene Details
Entrez Gene ID: | 643418 |
Gene Name: | lipase family member N |
Gene Aliases: |
ARCI8, LI4, LIPL4, bA186O14.3 |
Location: |
Chr.10:88757226-88783662 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LIPN | NM_001102469.1 | NP_001095939.1 | ||
| XM_005270049.3 | XP_005270106.1 | |||
| XM_011540083.2 | XP_011538385.1 | |||
| XM_011540084.2 | XP_011538386.1 | |||
| XM_011540085.2 | XP_011538387.1 | |||
| XM_011540086.2 | XP_011538388.1 | |||
| XM_017016549.1 | XP_016872038.1 | |||
| EF426483.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv551840 | Chr.10:88718726 - 88841108 on Build GRCh38 | Gain |
|
| nsv1043015 | Chr.10:88707310 - 88833935 on Build GRCh38 | Gain |
|
| nsv831941 | Chr.10:88650731 - 88827575 on Build GRCh38 | Gain |
|
| nsv825502 | Chr.10:88759237 - 88768756 on Build GRCh38 | Loss |
|
| esv3624139 | Chr.10:88724980 - 89003307 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map