Assay Details
Target Gene Details
Entrez Gene ID: | 8453 |
Gene Name: | cullin 2 |
Gene Aliases: |
- |
Location: |
Chr.10:35008551-35126899 on Build GRCh38 |
Assay Gene Location: | Within Exon 29 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CUL2 | NM_001198777.1 | 21 | 3251 | NP_001185706.1 |
| NM_001198778.1 | 21 | 3177 | NP_001185707.1 | |
| NM_001198779.1 | 21 | 3290 | NP_001185708.1 | |
| NM_001324375.1 | 19 | 3054 | NP_001311304.1 | |
| NM_001324376.1 | 22 | 3411 | NP_001311305.1 | |
| NM_003591.3 | 21 | 3243 | NP_003582.2 | |
| XM_011519743.1 | 23 | 3405 | XP_011518045.1 | |
| XM_011519744.1 | 22 | 3287 | XP_011518046.1 | |
| XM_011519745.1 | 22 | 3339 | XP_011518047.1 | |
| XM_011519747.1 | 22 | 3324 | XP_011518049.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv550401 | Chr.10:34931247 - 35060646 on Build GRCh38 | Gain |
|
| nsv550391 | Chr.10:34383535 - 35056041 on Build GRCh38 | Gain |
|
| dgv705n100 | Chr.10:34871114 - 35045560 on Build GRCh38 | Gain |
|
| dgv202e212 | Chr.10:34880967 - 35026718 on Build GRCh38 | Gain |
|
| nsv466869 | Chr.10:34966937 - 35052373 on Build GRCh38 | Gain |
|
| dgv706n100 | Chr.10:34881487 - 35230186 on Build GRCh38 | Gain |
|
| nsv550398 | Chr.10:34885707 - 35271661 on Build GRCh38 | Gain |
|
| nsv525060 | Chr.10:34786889 - 35020439 on Build GRCh38 | Gain |
|
| nsv1161937 | Chr.10:34971581 - 35049745 on Build GRCh38 | Duplication |
|
| dgv703n100 | Chr.10:34729183 - 35050396 on Build GRCh38 | Gain |
|
| nsv466862 | Chr.10:34695956 - 35011695 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map