Assay Details
Target Gene Details
Entrez Gene ID: | 4477 |
Gene Name: | microseminoprotein beta |
Gene Aliases: |
HPC13, IGBF, MSP, MSPB, PN44, PRPS, PSP, PSP-94, PSP57, PSP94 |
Location: |
Chr.10:46033305-46046269 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MSMB | NM_002443.3 | NP_002434.1 | ||
| NM_138634.2 | NP_619540.1 | |||
| AK312108.1 | ||||
| BC005257.1 | AAH05257.1 | |||
| BF680662.1 | ||||
| BM856408.1 | ||||
| S67815.1 | AAB29732.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1041519 | Chr.10:45991149 - 46080365 on Build GRCh38 | Gain |
|
| esv3577172 | Chr.10:45959639 - 46146408 on Build GRCh38 | Gain |
|
| nsv1042912 | Chr.10:46009162 - 46116183 on Build GRCh38 | Loss |
|
| nsv508583 | Chr.10:45881821 - 46146976 on Build GRCh38 | Deletion |
|
| esv3623185 | Chr.10:45995694 - 46106028 on Build GRCh38 | Gain |
|
| nsv550853 | Chr.10:46001360 - 46085061 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map