Assay Details
Target Gene Details
Entrez Gene ID: | 11101 |
Gene Name: | arginyltransferase 1 |
Gene Aliases: |
- |
Location: |
Chr.10:121740421-121928801 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATE1 | NM_001288736.1 | NP_001275665.1 | ||
| NR_110006.1 | ||||
| XM_005269459.3 | XP_005269516.1 | |||
| XM_017015598.1 | XP_016871087.1 | |||
| AK124669.1 |
Target Gene Details
Entrez Gene ID: | 100130887 |
Gene Name: | ATE1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.10:121928312-121951965 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATE1-AS1 | NR_120495.1 | |||
| AI634505.1 | ||||
| BF434328.1 |
More Information
Set Membership: |
|

Genomic Map